Chiller Supplies for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
G1833-67024
AGG1833-67024EA 248 USD
AGG1833-67024
Chiller Supplies for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Poly-clear fluid, strainer, and water filters for the ICP-OES G3292A chiller.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise.
Ensure that the ICP-OES G3292A chiller is optimally maintained by using the recommended chiller fluid and supplies. Poly-clear plus fluid is comprised of distilled water with algicide and rust inhibitor allowing for optimal chiller performanceMore Product Information