Heptane (mixture of isomers) ≥99% (n-heptane), Supelco®

Supplier: EMD MILLIPORE - AA – CORP

HX0080A-20
EM-HX0080A-20EA 11673 USD
EM-HX0080A-20
Heptane (mixture of isomers) ≥99% (n-heptane), Supelco®
Heptane (mixture of isomers)
Formula: H₃C(CH₂)₅CH₃
MW: 100.2 g/mol
Melting Pt: -90.5
Flash Pt: -4
MDL Number: MFCD00009544
CAS Number: 142-82-5
Merck Index: 13,04679

Order Now

Specification Test Results

Assay ≥99.9% (saturated C7 hydrocarbons)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR