Titanium ≥98%, fine powder, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Danger


MQ200

1.12379.0250
EM1.12379.0250CS 1608 USD
EM1.12379.0250
Titanium ≥98%, fine powder, Sigma-Aldrich®
Titanium
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
MDL Number: MFCD00011264
CAS Number: 7440-32-6
UN: 2546
ADR: 4.2
Merck Index: 12,09610

Order Now

Specification Test Results

Solubility (20 °C) insoluble,(reaction)
Melting Point 1670 °C
Molar Mass 47.88 g/mol
Bulk Density 1260 kg/m3
Boiling Point 3260 °C
Density 4.5 g/cm3 (20 °C)
pH 5 - 8 (50 g/l, H2O) suspension
Ignition Point 300 - 450 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR