EMPIGEN® BB Detergent, 30% Solution, Calbiochem®
Supplier: EMD MILLIPORE (BIOSCIENCES)
324690-100ML
EM324690-100MLEA 199.14 USD InStock
EM324690-100ML
EMPIGEN® BB Detergent, 30% Solution, Calbiochem®
Cleaning Agents Decontaminants
Lauryldimethylbetaine is useful as a detergent for solubilization of membrane components and for preparation of viral antigens.
  • A zwitterionic detergent
  • Useful for solubilization of keratins
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty