Ficoll 400, MP Biomedicals
Supplier: MP Biomedicals
0216000305 0216000310 0216000325 0216000380
IC16000305EA 98.38 USD
IC16000305 IC16000310 IC16000325 IC16000380
Ficoll 400, MP Biomedicals
Cell Separation Media
Ficoll® is a synthetic, hydrophilic polymer made by the copolymerisation of sucrose and epichlorhydrin. It is a highly branched polymer. Solutions at neutral pH can be sterilised by autoclaving.
  • MW: 400000 g/mol
FICOLL® has the follwing key advantages: Proven medium for density gradient centrifugation. It does not penetrate biological membranes and has high density solution of low viscosity and low osmotic pressure. Ir preserves functional and morphological integrity. It is useful for separating cells which are sensitive to centrifugation and forMore Product Information