LymphoSep™ Lymphocyte Separation Medium, MP Biomedicals
Supplier: MP Biomedicals
091692254
IC1692254EA 270.45 USD InStock
IC1692254
LymphoSep™ Lymphocyte Separation Medium, MP Biomedicals
Cell Separation Media
LymphoSep- Lymphocyte separation medium is a sterile, iso-osmotic, low viscosity solution for density centrifugation yielding a clear separation of lymphocytes from heparinized or defibrinated human blood.
  • In vitro isolation of lymphocytes from diluted whole blood
  • Maximum yield of mononuclear cells
  • Easy one-step centrifugation
  • >96% cell viability of lymphocytes
  • Low endotoxins
  • Autoclave sterilization
Ready to use quick, simple and reliable medium for the isolation of lymphocytes from whole blood. Lymphocytes are isolated by layering diluted,More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty