1,3-Dielaidin ≥99%, liquid

Supplier: E. Becker & Co. GmbH

0219515180 0219515190
IC19515180EA 149.49 USD
IC19515180 IC19515190
1,3-Dielaidin ≥99%, liquid
1,3-Dielaidin
Formula: C₃₉H₇₂O₅
MW: 621 g/mol
MDL Number: MFCD00042913
CAS Number: 25637-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR