Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Masterflex® Ismatec® Reglo Digital Piston Pump Drive, Avantor®

Supplier: MASTERFLEX LLC
78018-60 78018-60 78018-60 78018-60 78018-60 78018-60 78018-60 78018-60 78018-60
MFLX78018-60CS 0 USD InStock
MFLX78018-60 MFLX78018-60-AU MFLX78018-60-DK MFLX78018-60-IN MFLX78018-60-IL MFLX78018-60-EU MFLX78018-60-UK MFLX78018-60-CH MFLX78018-60-IT
Masterflex® Ismatec® Reglo Digital Piston Pump Drive, Avantor®
Pumps Pump Drives Piston Pump Drives
Achieve high pressure, accuracy, and repeatability -- with an intuitive touch-screen interface.

  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • Compatible with FMI RH series piston pump heads for flow ranges from 0.025 to 180 ml/min
  • 5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or analog remote control
  • Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
  • Pump lets you save and
More Product Information
Order Now
 

ORDER


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
US Plug Models
Collapse All
Description Plug type Max. Motor RPM Height Length Width Power Supplier No. VWR Catalog Number Unit Price Quantity
Ismatec® Reglo Digital Piston Pump with MasterflexLive®, 1800 rpm, 115 to 230 VAC US 1800 rpm 16 cm (6.3") 19.4 cm (7.6") 14.6 cm (5.8") 90 - 260 VAC; 50/60 Hz 78018-60 MFLX78018-60
  • Details

  • Each
    Retrieving
    2 year warranty MasterflexLive®
    Inquire for Price
    Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
    Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
    This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
    -Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
    This product has been blocked by your organization. Please contact your purchasing department for more information.
    The original product is no longer available. The replacement shown is available.
    This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
    Non-US Plug Models
    Collapse All
    Description Plug type Max. Motor RPM Height Length Width Power Supplier No. VWR Catalog Number Unit Price Quantity
    Ismatec® Reglo Digital Piston Pump with MasterflexLive®, 1800 rpm, 115 to 230 VAC AU 1800 rpm 16 cm (6.3") 19.4 cm (7.6") 14.6 cm (5.8") 90 - 260 VAC; 50/60 Hz 78018-60 MFLX78018-60-AU
  • Details

  • Each
    Retrieving
    2 year warranty MasterflexLive®
    Ismatec® Reglo Digital Piston Pump with MasterflexLive®, 1800 rpm, 115 to 230 VAC CH 1800 rpm 16 cm (6.3") 19.4 cm (7.6") 14.6 cm (5.8") 90 - 260 VAC; 50/60 Hz 78018-60 MFLX78018-60-CH
  • Details

  • Each
    Retrieving
    2 year warranty MasterflexLive®
    Ismatec® Reglo Digital Piston Pump with MasterflexLive®, 1800 rpm, 115 to 230 VAC DK 1800 rpm 16 cm (6.3") 19.4 cm (7.6") 14.6 cm (5.8") 90 - 260 VAC; 50/60 Hz 78018-60 MFLX78018-60-DK
  • Details

  • Each
    Retrieving
    2 year warranty MasterflexLive®
    Ismatec® Reglo Digital Piston Pump with MasterflexLive®, 1800 rpm, 115 to 230 VAC EU 1800 rpm 16 cm (6.3") 19.4 cm (7.6") 14.6 cm (5.8") 90 - 260 VAC; 50/60 Hz 78018-60 MFLX78018-60-EU
  • Details

  • Each
    Retrieving
    2 year warranty MasterflexLive®
    Ismatec® Reglo Digital Piston Pump with MasterflexLive®, 1800 rpm, 115 to 230 VAC IL 1800 rpm 16 cm (6.3") 19.4 cm (7.6") 14.6 cm (5.8") 90 - 260 VAC; 50/60 Hz 78018-60 MFLX78018-60-IL
  • Details

  • Each
    Retrieving
    2 year warranty MasterflexLive®
    Ismatec® Reglo Digital Piston Pump with MasterflexLive®, 1800 rpm, 115 to 230 VAC IN 1800 rpm 16 cm (6.3") 19.4 cm (7.6") 14.6 cm (5.8") 90 - 260 VAC; 50/60 Hz 78018-60 MFLX78018-60-IN
  • Details

  • Each
    Retrieving
    2 year warranty MasterflexLive®
    Ismatec® Reglo Digital Piston Pump with MasterflexLive®, 1800 rpm, 115 to 230 VAC IT 1800 rpm 16 cm (6.3") 19.4 cm (7.6") 14.6 cm (5.8") 90 - 260 VAC; 50/60 Hz 78018-60 MFLX78018-60-IT
  • Details

  • Each
    Retrieving
    2 year warranty MasterflexLive®
    Ismatec® Reglo Digital Piston Pump with MasterflexLive®, 1800 rpm, 115 to 230 VAC 5000 1800 rpm 16 cm (6.3") 19.4 cm (7.6") 14.6 cm (5.8") 90 - 260 VAC; 50/60 Hz 78018-60 MFLX78018-60-UK
  • Details

  • Each
    Retrieving
    2 year warranty MasterflexLive®
    Generate Barcodes Barcode Label Format: 
    Generate PDF Catalog Page

    Enter Details

    Select the article for which you are looking for Certificate

    Indicate the Lot Number. This field is mandatory.

    RELATED INFORMATION

    Enter Certificate Details

    Enter the Lot number:

    Choose from recent batch/lot numbers:

    Or
    Review & Compare Alternatives
    We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

    Original Product:

    Description Catalog Number Availability Unit Your Price Price Per Qty