Supplier: VWR International
MFLX98261-10 MFLX98261-20 MFLX98261-30
MFLX98261-10EA 9 USD InStock
MFLX98261-10 MFLX98261-20 MFLX98261-30
Stopcocks
Easily adjust flow.
  • Miniature plastic stopcocks have a straight bore
  • Ideal for liquids and gases under low pressure (operating pressure 7.25 psi/0.5 bar) or low vacuum.
The low friction properties of high density polyethylene in these miniature plastic stopcocks allows for easy turning of PP key which requires no lubricant, thus eliminating blockage of the bores with grease.
Order Now
ORDER
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty