EasySep™ Human CD33 Positive Selection Kit II
Supplier: Stemcell Technologies
17876
MSPP-17876EA 1280 USD InStock
MSPP-17876
EasySep™ Human CD33 Positive Selection Kit II
Cell Separation Media
Immunomagnetic positive selection of human CD33+ cells (e.g. myeloid cells).
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
  • Compatible across EasySep™, 'The Big Easy', and RoboSep™-S platforms
The EasySep™ Human CD33 Positive Selection Kit II is designed to isolate CD33+ cells from ammonium chloride lysed blood or bone marrow, or from nucleated cells prepared by density separation with RosetteSep™ DM-M density medium by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD33 and dextran-coatedMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty