EasySep™ Direct Human Pan-Granulocyte Isolation Kit
Supplier: Stemcell Technologies
19659
MSPP-19659EA 872.31 USD
MSPP-19659
EasySep™ Direct Human Pan-Granulocyte Isolation Kit
Cell Separation Media
Immunomagnetic isolation of human granulocytes directly from whole blood
  • >99.9% RBC depletion without the need for density gradient centrifugation, sedimentation or lysis
  • Up to 99% purity of isolated cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
The EasySep™ Direct Human Pan-Granulocyte Isolation Kit isolates functional, highly purified neutrophils, eosinophils, and basophils directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation, or other processing steps are requiredMore Product Information