EasySep™ Mouse Naïve CD8+ T Cell Isolation Kits
Supplier: Stemcell Technologies
19858
MSPP-19858EA 1349.23 USD InStock
MSPP-19858
EasySep™ Mouse Naïve CD8+ T Cell Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched mouse naïve CD8+ T cells
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse Naïve CD8+ T Cell Isolation Kit is designed to isolate naïve CD62L+CD44-CD8+ T cells from single-cell suspensions of splenocytes or lymph nodes by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-naïve CD8+ T cells (CD4, CD11b, CD11c, CD19, CD44, CD45R/B220, CD49b, TCRγ/δ, TER119) and streptavidin-coated magneticMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty