HPLC Column Heater, Cole Palmer
Supplier: Cole Parmer
42650-70
MSPP-UX4265070EA 1589.75 USD
MSPP-UX4265070
HPLC Column Heater, Cole Palmer
Heaters Column Heaters
Suitable for the columns that are 1/4" or 3/8" diameter by up to 30 cm long.
  • Compact
  • Easy-to use
  • Features up and down arrow for temperature set-up
  • With 30-days count down timer
  • Features platinum RTD temperature sensor and PID temperature controller
The silicon rubber heater underneath the chamber provides uniform chamber temperatures for optimal results. The chamber design provides quick access to the column and features a clear polycarbonate cover for easy viewing. Target and actual temperatures will be displayed—the stable temperature light willMore Product Information