Restore™ Western Blot Stripping Buffer, Thermo Scientific
Supplier: Paul Schöndorf Metallwaren GmbH
21059 21062 21063
PI21059EA 212.15 USD
PI21059 PI21062 PI21063
Restore™ Western Blot Stripping Buffer, Thermo Scientific
Buffers Electrophoresis Buffers Stripping Buffers
Thermo Scientific Restore™ Western Blot Stripping Buffer safely and effectively removes primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.
  • Saves time—no need to re-run gels and blots
  • Saves costly sample—re-probe the membrane using the same target sample
  • Effective—formulation is more efficient at stripping antibodies than homemade buffers
  • Gentle—does not damage the target antigen during stripping allowing efficient reprobing
  • Odor-free—no mercaptans means no acrid odors
  • Economical—less expensive than other commercial stripping buffers
Performing gel electrophoresis and duplicate immunoblot assays to test new primary antibodies or antibody concentrations is time-consuming and expensive. Restore™ Western blot stripping buffer offers an effective alternative.
Precaución: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store at room temperature or 4˚C. Product shipped at ambient temperature.