Ammonium dihydrogen phosphate 20,000 ppm in nitric acid 1%, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy

Supplier: RICCA CHEMICAL CO
Danger

RV010137-50N
RCRV010137-50NEA 269 USD
RCRV010137-50N RCRV010137100N
Ammonium dihydrogen phosphate 20,000 ppm in nitric acid 1%, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy
Ammonium dihydrogen phosphate

Certificaciones: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
Merck Index: 13,00547

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR