Dichloromethylvinylsilane 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC14743-5000
TS14743-5000EA 125 USD
TS14743-5000
Dichloromethylvinylsilane 97%
Dichloromethylvinylsilane
Formula: C₃H₆Cl₂Si
MW: 141.07 g/mol
Boiling Pt: 92 °C (760 mmHg)
Density: 1.08 g/cm³
Flash Pt: 4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000492
CAS Number: 124-70-9
EINECS: 204-710-3
UN: 2985
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4310 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR