SSC Buffer, Saline-Sodium Citrate, 20X Solution, Thermo Scientific
Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
61152-0010
TS61152-0010EA 144 USD
TS61152-0010
SSC Buffer, Saline-Sodium Citrate, 20X Solution, Thermo Scientific
Buffers Electrophoresis Buffers Hybridization Buffers
DNase-, RNase- and protease-free.
MDL No.: MFCD00132371.