|
![]() Analytical Sample Prep
![]() The Next Generation Syringe FilterThe new Acrodisc One™ is the next generation syringe filter for aqueous and aggressive organic solvent-based solutions. Extend column life up to 52 times with up to 24% less filter extractables than the competition. ![]() Simplify Mobile Phase Filtration with SolVac®Simplify Clean-up and Degassing of Mobile Phase Solvents. With a versatile, durable and leak-proof design, draw directly from HPLC Solvent bottles for less spill risk, breakage, and minimal risk of contamination. ![]() Acrodisc MS certified Syringe FiltersThe Acrodisc MS is the first syringe filter that is LCMS-certified. The filters are designed for and certified to be very low in LCMS extractables providing more accurate results to the end user. ![]() Ion Chromatography (IC) Acrodisc Syringe FiltersPall’s Acrodisc Ion Chromatography (IC) syringe filters have been tested to monitor for inorganic extractables. For ion chromatograph applications, only the Acrodisc IC (PES) syringe filter is certified for low levels of inorganic extractables. |
![]() Analytical Sample Prep
|