5-Chloro-1-pentanol 90,0
Supplier: Decon Laboratories Ltd
C0815-25ML
C0815-5ML
TCC0815-25MLEA
140
USD
TCC0815-25ML
TCC0815-5ML
5-Chloro-1-pentanol 90,0
5-Chloro-1-pentanol
CAS Number: 5259-98-3
MDL Number: MFCD00039556
Molecular Formula: C5H11ClO
Molecular Weight: 122.59
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Specific Gravity (20/20): 1.06
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Pentamethylene Chlorohydrin
Formula:
C₅H₁₁ClO MW: 122.59 g/mol Boiling Pt: 99 °C Density: 1.06 (20 °C) |
MDL Number:
MFCD00039556 CAS Number: 5259-98-3 |
|
Specification Test Results
Purity/Analysis Method | >90.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...