Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Diaminotoluene ≥97.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

D3788-25G D3788-5G
TCD3788-25GEA 655 USD InStock
TCD3788-25G TCD3788-5G
2,3-Diaminotoluene ≥97.0% (by GC, titration analysis)
2,3-Diaminotoluene
CAS Number: 2687-25-4
MDL Number: MFCD00011589
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 225
Melting point (°C): 60

Other Product Names / Synonyms: 3-Methyl-1,2-phenylenediamine, Tolylene-2,3-diamine


Formula: C₇H₁₀N₂
Boiling Pt: 225 °C
Melting Pt: 60 °C
MDL Number: MFCD00011589
CAS Number: 2687-25-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b739e1facb29f5bee30ea0faf10ad0ece4001e0&vid=8b739e1facb29f5bee30ea0faf10ad0ece4001e0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty