Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethyl-2-phenylindole ≥98.0%

Supplier: Decon Laboratories Ltd

E0331-25G E0331-500G
TCE0331-025GEA 28 USD InStock
TCE0331-025G TCE0331-500G
1-Ethyl-2-phenylindole ≥98.0%
1-Ethyl-2-phenylindole
CAS Number: 13228-39-2
MDL Number: MFCD00031481
Molecular Formula: C16H15N
Molecular Weight: 221.30
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Boiling point (°C): 209
Melting point (°C): 86


Formula: C₁₆H₁₅N
MW: 221.30 g/mol
Boiling Pt: 209 °C
Melting Pt: 86 °C
MDL Number: MFCD00031481
CAS Number: 13228-39-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea7566d5c1b4a5b9ab9910a9e24714cd8fb38577&vid=ea7566d5c1b4a5b9ab9910a9e24714cd8fb38577&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty