I0745-25G
I0745-5G
TCI0745-25GEA
108.85
USD
InStock
TCI0745-25G
TCI0745-5G
1-Isopropylpiperazine ≥98.0%
1-Isopropylpiperazine
CAS Number: 4318-42-7
MDL Number: MFCD00167971
Molecular Formula: C7H16N2
Molecular Weight: 128.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 178
Flash Point (°C): 54
Specific Gravity (20/20): 0.90