Sulfur trioxide pyridine complex 95,0

Supplier: Decon Laboratories Ltd

P0998-500G P0998-25G
TCP0998-500GEA 679 USD
TCP0998-500G TCP0998-25G
Sulfur trioxide pyridine complex 95,0
Sulphur trioxide pyridine complex

CAS Number: 26412-87-3
MDL Number: MFCD00012437
Molecular Formula: C5H5N
Molecular Weight: 159.16
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Others (Sulfur Compounds for Synthesis), Oxidation
Other Product Names / Synonyms: Sulfur Trioxide - Pyridine Complex

Formula: C₅H₅NO₃S
MW: 159.16 g/mol
MDL Number: MFCD00012437
CAS Number: 26412-87-3
UN: 3261
ADR: 8 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f352e776f8c179952ef6aa47be38ce9dbf97a297&vid=f352e776f8c179952ef6aa47be38ce9dbf97a297&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR