Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Tyrosine methyl ester hydrochloride ≥98.0%

Supplier: Decon Laboratories Ltd

T1108-5G T1108-25G
TCT1108-005GEA 22 USD InStock
TCT1108-005G TCT1108-025G
L-Tyrosine methyl ester hydrochloride ≥98.0%
L-Tyrosine methyl ester hydrochloride
CAS Number: 3417-91-2
MDL Number: MFCD00012607
Molecular Formula: C10H13NO3
Molecular Weight: 231.68
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: 12 deg (C=2, MeOH)

Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Tyr-OMe.HCl


Formula: C₁₀H₁₃NO₃
MW: 231.68 g/mol
MDL Number: MFCD00012607
CAS Number: 3417-91-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f98273b9f6074389e8dabb02eb7b670f2c459474&vid=f98273b9f6074389e8dabb02eb7b670f2c459474&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty