Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc 2-methoxyethoxide 5% (w/v) in 2-methoxyethanol

Supplier: Origen Biomedical, Inc
Danger

043903-AE 043903-AC
AAA43903-AEEA 462 USD InStock
AAA43903-AE AAA43903-AC
Zinc 2-methoxyethoxide 5% (w/v) in 2-methoxyethanol
Zinc 2-methoxyethoxide
Liquid

CAS No.: 129918-15-6 Molecular Formula: C6H14O2Zn Formula Weight: 215.56 Flash Point: 40°(104°F) Physical Form: Liquid Density: 0.96 Sensitivity: Moisture Sensitive Appearance: Pale yellow Boiling Point: 124° UN No.: 1188 MDL No.: MFCD00156529


Formula: C₆H₁₄O₂Zn
MW: 215.56 g/mol
Boiling Pt: 124 °C
Density: 0.96
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00156529
CAS Number: 129918-15-6
UN: 1188
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty