Polyester acrylate/methacrylate PEAM-645

Supplier: Origen Biomedical, Inc
Warning

046596-14 046596-22
AA46596-14EA 80.1 USD
AA46596-14 AA46596-22
Polyester acrylate/methacrylate PEAM-645
Polyester acrylate/methacrylate

Amber liquid


Applications: Recommended for use as a base resin in adhesive applications or coating applications


Applications: Recommended for use as a base resin in adhesive applications or coating applications

MW: 645 g/mol
Melting Pt: >375 °C (dec.)
Storage Temperature: Refrigerator
CAS Number: 921213-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR