2-Amino-5-bromopyrazine 97%
Supplier: Origen Biomedical, Inc
|
Warning
|
H27465-03
H27465-06
AAH27465-03EA
163
USD
AAH27465-03
AAH27465-06
2-Amino-5-bromopyrazine 97%
2-Amino-5-bromopyrazine
Purity: 97%
CAS No.: 59489-71-3
Molecular Formula: C4H4BrN3
Formula Weight: 174.00
Melting Point: 113-117°
MDL No.: MFCD00235015
Formula:
C₄H₄BrN₃ MW: 174.00 g/mol Melting Pt: 113…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00235015 CAS Number: 59489-71-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...