3-(tert-Butyldimethylsilyloxy)phenylboronic acid tech. 90%, tech.

Supplier: Origen Biomedical, Inc
Warning

H28378-03 H28378-06
AAH28378-03EA 68.7 USD
AAH28378-03 AAH28378-06
3-(tert-Butyldimethylsilyloxy)phenylboronic acid tech. 90%, tech.
3-(tert-Butyldimethylsilyloxy)phenylboronic acid

Purity: 90% CAS No.: 261621-12-9 Molecular Formula: C12H21BO3Si Formula Weight: 252.19 Melting Point: 86-91° MDL No.: MFCD03701505

Formula: C₁₂H₂₁BO₃Si
MW: 252.19 g/mol
Melting Pt: 86…91 °C
Storage Temperature: Ambient
MDL Number: MFCD03701505
CAS Number: 261621-12-9