4-Methoxy-trans-β-styrylboronic acid pinacol ester 98%

Supplier: Origen Biomedical, Inc

H30373-MD H30373-03 H30373-06
AAH30373-MDEA 68.6 USD
AAH30373-MD AAH30373-03 AAH30373-06
4-Methoxy-trans-β-styrylboronic acid pinacol ester 98%
4-Methoxy-trans-beta-styrylboronic acid pinacol ester

Purity: 98% CAS No.: 149777-83-3 Molecular Formula: C15H21BO3 Formula Weight: 260.14 MDL No.: MFCD04038966

Formula: C₁₅H₂₁BO₃
MW: 260.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038966
CAS Number: 149777-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR