4-(Dimethylamino)benzoic anhydride 97%
Supplier: Origen Biomedical, Inc
|
Warning
|
H32503-03
H32503-06
AAH32503-03EA
50.4
USD
AAH32503-03
AAH32503-06
4-(Dimethylamino)benzoic anhydride 97%
4-(Dimethylamino)benzoic anhydride
Purity: 97%
CAS No.: 7474-31-9
Molecular Formula: C18H20N2O3
Formula Weight: 312.37
Melting Point: 149-154°
MDL No.: MFCD09757554
Formula:
C₁₈H₂₀N₂O₃ MW: 312.37 g/mol Melting Pt: 149…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD09757554 CAS Number: 7474-31-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...