H33795-03
H33795-06
AAH33795-03EA
84.9
USD
InStock
AAH33795-03
AAH33795-06
3-(1-Naphthyl)propionic acid 96%
3-(1-Naphthyl)propionic acid
Purity: 96%
CAS No.: 3243-42-3
Molecular Formula: C13H12O2
Formula Weight: 200.24
Melting Point: 155-159°
MDL No.: MFCD00033058