Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)

Supplier: Origen Biomedical, Inc
Danger

H51170-AE H51170-AP
AAH51170-AEEA 130 USD InStock
AAH51170-AE AAH51170-AP
Allylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)
Allylmagnesium chloride
CAS No.: 2622-05-1 Molecular Formula: C3H5ClMg Formula Weight: 100.83 UN No.: 3399 MDL No.: MFCD00000473


Formula: C₃H₅ClMg
Storage Temperature: Ambient
MDL Number: MFCD00000473
CAS Number: 2622-05-1
UN: 3399
ADR: 4.3,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty