Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-3,3-Dimethyl-1-butenylboronic acid 97%

Supplier: Origen Biomedical, Inc
Warning

H52627-03 H52627-MD
AAH52627-03EA 245 USD InStock
AAH52627-03 AAH52627-MD
trans-3,3-Dimethyl-1-butenylboronic acid 97%
trans-3,3-Dimethyl-1-butenylboronic acid
Purity: 97% CAS No.: 86595-37-1 Molecular Formula: C6H13BO2 Formula Weight: 127.98 Melting Point: 78-80° MDL No.: MFCD01074624


Formula: C₆H₁₃BO₂
MW: 127.98 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD01074624
CAS Number: 86595-37-1
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty