H53010-03
H53010-MD
AAH53010-03EA
132
USD
InStock
AAH53010-03
AAH53010-MD
5-Carbamoyl-2-fluorophenylboronic acid 95%
5-Carbamoyl-2-fluorophenylboronic acid
Purity: 95%
CAS No.: 874289-39-1
Molecular Formula: C7H7BFNO3
Formula Weight: 182.95
Melting Point: 192-194°
MDL No.: MFCD08436055