H53227-03
H53227-06
AAH53227-03EA
71.8
USD
InStock
AAH53227-03
AAH53227-06
E-Phenylethenylboronic acid 97%
E-Phenylethenylboronic acid
Purity: 97%
CAS No.: 6783-05-7
Molecular Formula: C8H9BO2
Formula Weight: 147.97
Storage Temperature: -30°C to -10°C
Melting Point: 98-100°
MDL No.: MFCD00963621