H53455-03
H53455-06
AAH53455-03EA
102
USD
InStock
AAH53455-03
AAH53455-06
4-Ethynylpyridine hydrochloride 97%
4-Ethynylpyridine hydrochloride
Purity: 97%
CAS No.: 352530-29-1
Molecular Formula: C7H5N.HCl
Formula Weight: 139.58
Melting Point: ca 150° dec.
MDL No.: MFCD03093912