H55470-06
H55470-14
AAH55470-03EA
31.62
USD
InStock
AAH55470-03
AAH55470-06
AAH55470-MD
AAH55470-14
Ethyl-O-Mesitylsulfonylacetohydroxamate ≥98%
Ethyl-O-Mesitylsulfonylacetohydroxamate
Purity: 98+%
CAS No.: 38202-27-6
Molecular Formula: C13H19NO4S
Formula Weight: 285.36
Storage Temperature: 2°C to 8°C
Melting Point: 54-56°
Beilstein Reference: 2144156
MDL No.: MFCD00009244