Sodium cacodylate, 0.1 M buffer soln., pH 6.5, Thermo Scientific

Supplier: Origen Biomedical, Inc

J60367-AE J60367-AD
AAJ60367-AEEA 152.11 USD
AAJ60367-AE AAJ60367-AD
Sodium cacodylate, 0.1 M buffer soln., pH 6.5, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 6131-99-3
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
MDL No.: MFCD00149079

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR