Sodium Phosphate, 0.2 M Buffer Solution, pH 8.0, Thermo Scientific
Supplier: Origen Biomedical, Inc
J62733-AP
J62733-K2
AAJ62733-APEA
173.96
USD
AAJ62733-AP
AAJ62733-K2
Sodium Phosphate, 0.2 M Buffer Solution, pH 8.0, Thermo Scientific
Buffers
Biological Buffers
Liquid
Physical Form: Liquid
MDL No.: MFCD00146206
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...