L14053-03
L14053-06
L14053-14
AAL14053-03EA
39.1
USD
InStock
AAL14053-03
AAL14053-06
AAL14053-14
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol 99%
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol
Purity: 99%
CAS No.: 18680-27-8
Molecular Formula: C10H18O2
Formula Weight: 170.25
Storage Temperature: 2°C to 8°C
Flash Point: 110°(230°F)
Melting Point: 56-58°
Beilstein Reference: 1853591
Optical Rotation: +8.7° (c=6.5 in toluene)
MDL No.: MFCD00077851