L20448-0B
L20448-22
L20448-30
AAL20448-0BEA
155
USD
InStock
AAL20448-0B
AAL20448-22
AAL20448-30
Cyanamide 50% (w/w) in aqueous solution stabilized
Cyanamide
CAS No.: 420-04-2
Molecular Formula: CH2N2
Formula Weight: 42.04
Storage Temperature: 2°C to 8°C
Flash Point: None
Refractive Index: 1.3980
Odor: Odorless
pH: 4.2
Beilstein Reference: 1732569
Merck Reference: 14,2684
UN No.: 2922
MDL No.: MFCD00007572