D-(+)-Glucose, anhydrous
Supplier: THERMO SCIENTIFIC CHEMICALS INC MS
J14535-P5
J14535-Q1
J14535-Q5
101172-434EA
92.09
USD
101172-434
101170-420
101175-008
D-(+)-Glucose, anhydrous
D-(+)-Glucose
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Melting Pt: 146 Storage Temperature: Ambient |
MDL Number:
MFCD00148912 CAS Number: 50-99-7 EINECS: 200-075-1 Merck Index: 14,04459 |
|
Specification Test Results
Identity | By IR |
Specific Rotation (25°C) | +52.6 to 53.2° (dry basis, c=10 in 0.012 N NH₄OH) |
Moisture (KF) | ≤1.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...