Bovine;Dog;Human;Mouse;Pig;Rat Endothelin-1 Acetate
Supplier: Frontier Scientific
4040254.0001 4040254.0005
H-6995.0001BAEA 1334.25 USD InStock
H-6995.0001BA H-6995.0005BA
Bovine;Dog;Human;Mouse;Pig;Rat Endothelin-1 Acetate
Proteins and Peptides
Endothelins are peptides with exceptional vasoconstrictor potency. They play an important role in intercellular communications.
Endothelin-1, a 21-amino acid peptide from vascular endothelial cells of different mammalian species, is one of the most potent vasoconstrictors. ET-1 exerts a wide variety of effects on both vascular and non-vascular (e.g. heart, lung, brain) tissues. Toxic peptides from a snake venom, the sarafotoxins, show structural and functional homology to ET-1. The role of ET-1 in clinical hypertension has been reviewed by Dhaun et al. and by Sreenivas and Oparil. ET-1 plays a central role in lung fibrosis.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty