Anti-VAMP2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
VAS-SV006-D VAS-SV006-F
95046-464EA 232.17 USD
95046-464 95046-466
Anti-VAMP2 Rabbit Polyclonal Antibody
Antibodies
Vesicle-associated membrane protein (VAMP), also known as synaptobrevin, is an 18 kDa integral membrane protein localized to the cytoplasmic surface of synaptic vesicles. Two VAMP homologs, VAMP1 and VAMP2, are differentially expressed in the nervous system. VAMP2 is found in neurons associated with autonomic, sensory and integrative functions, and in non-neuronal Langerhans islets and glomerular cells. VAMP is a component of a 20S complex implicated in vesicle docking and fusion, the formation of which involves the interaction of VAMP with syntaxin through an ATP-dependent reaction modulated by synaptotagmin, SNAP-25, alpha-SNAP, and NSF.

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, guinea pig, and rabbit VAMP2. Detects a band of ~18kDa by Western blot.

Type: Primary
Antigen: VAMP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR