DPQ ≥99%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

AG-CR1-0037-M001 AG-CR1-0037-M005
102516-024EA 55.38 USD
102516-024 102516-026
DPQ ≥99%
DPQ

Very potent PARP-1 inhibitor.

Formula: C₁₈H₂₆N₂O₂
MW: 302.4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01866274
CAS Number: 129075-73-6

Order Now

Specification Test Results

Purity 99% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO; slightly soluble in 100% ethanol; insoluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR