Fumitremorgin C ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste
Danger

BVT-0189-C250 BVT-0189-M001
102513-580EA 117.69 USD
102513-580 102513-582
Fumitremorgin C ≥98%
Fumitremorgin C

Mycotoxin. Cytotoxic. Tremorgenic. Potent and specific inhibitor of the breast cancer resistance protein (BCRP; ABCG2). Reverses multidrug resistance mediated by BCRP and increases cytotoxicity of several anticancer agents in vitro.

Formula: C₂₂H₂₅N₃O₃
MW: 379.5 g/mol
Storage Temperature: Refrigerator
CAS Number: 118974-02-0
UN: 3462
ADR: 6.1 III

Order Now

Specification Test Results

Source/Host Isolated from Aspergillus fumigatus.
Purity 98% (HPLC)
Appearance/Color White to light brown powder.
Solubility Soluble in DMSO, methanol, acetone, DMF or chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR