Sodium borohydride, tablets for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Danger


M-Clarity™ quality level = MQ200

8.18823.0025 8.18823.0100
EM8.18823.0025CS 2073 USD
EM8.18823.0025 EM8.18823.0100
Sodium borohydride, tablets for synthesis, Sigma-Aldrich®
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400
Flash Pt: 69
MDL Number: MFCD00003518
CAS Number: 16940-66-2
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664

Order Now

Specification Test Results

Solubility 550 g/l (25 °C) (slow decomposition)
Melting Point 400 °C (slow decomposition)
Molar Mass 37.83 g/mol
Bulk Density 350 - 500 kg/m3
Boiling Point 500 °C (decomposition)
Explosion Limit 3.02 %(V)
Density 1.07 g/cm3 (20 °C)
pH 11 (10 g/l, H2O, 20 °C)
Ignition Point 220 °C
Hazard Class 4.3
Hazard Class Description Dangerous When Wet Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR