Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl fluoroacetate 98%

Supplier: Origen Biomedical, Inc
Danger

H61340-09 H61340-18
AAH61340-09EA 44.6 USD InStock
AAH61340-09 AAH61340-18
Ethyl fluoroacetate 98%
Ethyl fluoroacetate
Ethyl fluoroacetate, 98%


Formula: C₄H₇FO₂
Boiling Pt: 119…120 °C
Density: 1.0912 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000450
CAS Number: 459-72-3
EINECS: 207-297-8
UN: 2929
ADR: 6.1,II
Merck Index: 14,4167
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty