Water nuclease-free, sterile, DEPC treated

UPW-0125-4 UPW-0125-48 UPW-0500-2 UPW-0500-20 UPW-1000-12 UPW-2000-6
101076-142PK 82.35 USD
101076-142 101076-144 101076-146 101076-148 101076-150 101076-242
Water nuclease-free, sterile, DEPC treated
Water

DEPC treatment is an effective way to eliminate RNAse activity. Our DEPC-treated water is made using the least amount of DEPC that is still effective at destroying enzymatic activitiy in RNases and other proteins, making it ideal for RNA extraction and all molecular biology applications. Water is autoclaved both before and after packaging to ensure sterility and to complete inactivation of byproducts of DEPC. CDI and UV treated.


  • Certified DNase/RNase free


Growcells provides the highest quality of Nuclease Free water for critical applications. Water is purified, tested and certified to be devoid of any activiy in DNAse, RNAse, and Protease assays. Available in convenient volumes and packaging to fit all applications requiring biologically inactive water.
Polycarbonate bottle packaging is re-usable and autoclavable.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now

Specification Test Results

Recipe 0.01ml DEPC
Sterile Filtered Yes (0.10 μM)
Storage RT
Shipping Ground
Shelf Life 3 Years (from Date of Manufacture)
Sterility SAL 10-6
Endotoxins < 0.005eu/ml
Activity None Detected
Resistivity 18.2mΩ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR