Gray Silicone Septum, Ace Glass Incorporated
Supplier: Ace Glass
12904-06 12904-08 12904-10
89077-542PK 35.28 USD
89077-542 89077-544 89077-546
Gray Silicone Septum, Ace Glass Incorporated
Septa
Septa are flat 3 mm thick gray silicone
Also for use with Ace-Thred fittings.